Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell™ Seamless Knit Cotton/Polyester Gloves with PVC Web Grip

Catalog No. 19120807A
Change view
Click to view available options
Small
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
19-120-807A Small
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-120-807A Supplier Honeywell™ Supplier No. WB18GRS
This item is not returnable. View return policy

Seamless knit construction is comfortable and flexible, with PVC coating in a webbed design for enhanced grip capabilities.

  • Knit wrist helps prevent dirt and debris from entering the glove
TRUSTED_SUSTAINABILITY

Specifications

Construction Seamless Knit
Coating Material PVC
Color Gray
Hand Ambidextrous
Material Polycotton
Product Type Seamless Knit Gloves
Size Small
For Use With (Application) Automotive Assembly, Commercial Fishing, Manufacturing, and Warehousing
Glove Cuff Style Knit
Series WB18GR
Yarn Gauge 7
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.