Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell™ Lightweight PVC Coated Seamless Knit Cotton/Polyester Gloves

Catalog No. 19059928
Change view
Click to view available options
Large
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
19-059928 Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-059928 Supplier Honeywell™ Supplier No. NVC13A
This item is not returnable. View return policy

PVC coating on the palm provides abrasion resistance, and enhances grip capabilities.

  • Lightweight cotton/polyester gloves
  • Seamless construction minimizes irritation caused by rubbing or chaffing
  • PVC coating on the palm enhances grip capabilities
TRUSTED_SUSTAINABILITY

Specifications

Coated Area Palm
Construction Seamless Knit
Coating Material PVC
Color Natural, Red
Hand Specific
Material Polycotton
Product Type Seamless Knit Gloves
Size Large
For Use With (Application) Assembly, Parts Handling, Recycling, General Maintenance, and Warehousing
Gender Men's
Glove Cuff Color Natural
Glove Cuff Style Knit
Series NVC13
Yarn Gauge 13
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.