Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell™ POWERCOAT™ Neoprene Chemical Resistant Gloves

Catalog No. 191301772D
Change view
Click to view available options
Size:
Small
X-Large
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-130-1772D X-Large
19-130-1772A Small
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-130-1772D Supplier Honeywell™ Supplier No. PCNE14XL

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Neoprene gloves are 14 in. in length and have a multitude of resistant properties to oils, acids, caustics and solvents.

  • Neoprene has resistance to a broad range of oils, acids, caustics and solvents
  • Secure grip in greasy or oily applications
  • Flame retardent and resistance to sunlight and ozone degradation
  • Knit lining wicks away hand moisture
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ASTM 3389-05
Certifications/Compliance (Europe) Category III
Applications Industrial
Color Blue, Black
Latex Free Yes
Material Neoprene
Size X-Large
EN388 Standard EN 388:3121X
EN ISO 374-1 Type A
EN ISO 374-1 CHEMICAL LETTER CODES AKLMNOPT
Glove Cuff Style Pinked
Grip Design Rough
Liner Material Cotton Interlock
Lining Lined
Model PC-NE14-XL
Series POWERCOAT™
Length (English) 14 in.
Length (Metric) 35.5 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.