Learn More
Honeywell™ PRO 3009 Leather Boots
Catalog No. 19814364
Change view
Click to view available options
Size: 10D
Catalog No.
19-814-364 Size: 10D
1 options
This item is not returnable. View return policy
Catalog No. 19-814-364 Supplier Honeywell™ Supplier No. BT30091002
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.
This item is not returnable. View return policy
CrossTech™ fabric bootie system laminated to quilted, fungus-resistant Cambrelle™ and Kevlar™ felt linings
  • Corrosion-resistant steel ladder shank and puncture-resistant bottom plate
  • Double-stitched, reinforced aggressive lug soles
  • 14 in. high black, pull-on bunker style boot with side and back stays made from top-grade Military AB water-resistant leather
  • Oblique safety-toe cap
  • Slip-resistant tread design
  • Full wraparound Goodyear™ Welt
Compliance
Meet NFPA 1971, 2007 Edition requirements.
D width. Additional widths available through your Fisher Scientific™ Customer Service Representative.
TRUSTED_SUSTAINABILITY
Specifications
Height (English) 14 in.
Material Leather, CrossTech
Size 10D