Learn More
Honeywell™ PRO 5006 Structural Firefighting Boots, E Width
Catalog No. 19167427
Change view
Click to view available options
Material:
Leather, Dupont Kevlar Aramid Fiber, CrossTech
Pro-V Liner
Size:
10.5E
10E
11.5E
11E
12.5E
12E
13E
14E
15E
16E
5.5E
5E
6.5E
6E
7.5E
7E
8.5E
8E
9.5E
9E
20 product options available for selection
Product selection table with 20 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material Size
19-167-427 Leather, Dupont Kevlar Aramid Fiber, CrossTech 10.5E
19-167-426 Pro-V Liner 10E
19-167-429 Leather, Dupont Kevlar Aramid Fiber, CrossTech 11.5E
19-167-428 Pro-V Liner 11E
19-167-431 Leather, Dupont Kevlar Aramid Fiber, CrossTech 12.5E
19-167-430 Leather, Dupont Kevlar Aramid Fiber, CrossTech 12E
19-167-432 Pro-V Liner 13E
19-167-433 Pro-V Liner 14E
19-167-434 Leather, Dupont Kevlar Aramid Fiber, CrossTech 15E
19-167-435 Pro-V Liner 16E
19-167-417 Leather, Dupont Kevlar Aramid Fiber, CrossTech 5.5E
19-167-416 Pro-V Liner 5E
19-167-418 Pro-V Liner 6E
19-167-419 Leather, Dupont Kevlar Aramid Fiber, CrossTech 6.5E
19-167-421 Leather, Dupont Kevlar Aramid Fiber, CrossTech 7.5E
19-167-420 Pro-V Liner 7E
19-167-423 Leather, Dupont Kevlar Aramid Fiber, CrossTech 8.5E
19-167-422 Pro-V Liner 8E
19-167-424 Pro-V Liner 9E
19-167-425 Leather, Dupont Kevlar Aramid Fiber, CrossTech 9.5E
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 20 options available.
20 options
Catalog No. 19-167-427 Supplier Honeywell™ Supplier No. BT5006B1053
Add to Cart
Add to Cart
Superior combination of comfort and absorbency
Quilted to 7.5 oz Kevlar™ felt thermal barrier and laminated to fully encapsulating Crosstech™ Footwear Fabric bootie. Crosstech™ moisture barrier provides superior liquid penetration resistance to blood, body fluids and NFPA tested common chemicals.
  • Military AB leather construction insulates and adds abrasion-, puncture-, water- and chemical-resistance
  • With heavy duty 0.75 in. reinforced straps mounted to the exterior of boot and anchored with a 2 × 1.5 in. shield-shaped base
  • External texon board shinguard protects and guards sensitive shin area of legs
  • Patented PowerToe™ cover resists toe abrasion and provides unparalled wear over the lifetime of the boots
  • Oblique toe safety cap provides additional room in height and width of boot
  • Fire/Ice PowerSole™ angled lugs provide substantial edging and gripping capabilities
  • Molded thermoplastic heel counter locks foot into the boot and anchors it in place
  • Patented PowerHeel™ cover protects heel area from abusive doffing of boots
  • Corrison-resistant steel ladder shank and stainless steel, full length, toe-to-heel puncture resistant bottom plate
Order Info
D and EEE widths also available.
Compliance
Meet NFPA 1971, 2007 edition requirements
TRUSTED_SUSTAINABILITY
Specifications
Height (English) 14 in.
Material Leather, Dupont Kevlar Aramid Fiber, CrossTech
Size 10.5E