Learn More
Honeywell™ PRO 5006 Structural Firefighting Boots, EEE Width
Catalog No. 19167455
Change view
Click to view available options
Material:
Leather, Dupont Kevlar Aramid Fiber, CrossTech
Pro-V Liner
Size:
10.5EEE
10EEE
11.5EEE
11EEE
12.5EEE
12EEE
13EEE
14EEE
15EEE
16EEE
5.5EEE
6.5EEE
6EEE
7.5EEE
7EEE
8.5EEE
8EEE
9.5EEE
9EEE
19 product options available for selection
Product selection table with 19 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material Size
19-167-455 Leather, Dupont Kevlar Aramid Fiber, CrossTech 16EEE
19-167-447 Leather, Dupont Kevlar Aramid Fiber, CrossTech 10.5EEE
19-167-446 Leather, Dupont Kevlar Aramid Fiber, CrossTech 10EEE
19-167-451 Leather, Dupont Kevlar Aramid Fiber, CrossTech 12.5EEE
19-167-448 Leather, Dupont Kevlar Aramid Fiber, CrossTech 11EEE
19-167-450 Leather, Dupont Kevlar Aramid Fiber, CrossTech 12EEE
19-167-452 Leather, Dupont Kevlar Aramid Fiber, CrossTech 13EEE
19-167-453 Leather, Dupont Kevlar Aramid Fiber, CrossTech 14EEE
19-167-454 Leather, Dupont Kevlar Aramid Fiber, CrossTech 15EEE
19-167-437 Leather, Dupont Kevlar Aramid Fiber, CrossTech 5.5EEE
19-167-438 Pro-V Liner 6EEE
19-167-439 Leather, Dupont Kevlar Aramid Fiber, CrossTech 6.5EEE
19-167-440 Pro-V Liner 7EEE
19-167-441 Leather, Dupont Kevlar Aramid Fiber, CrossTech 7.5EEE
19-167-443 Leather, Dupont Kevlar Aramid Fiber, CrossTech 8.5EEE
19-167-442 Pro-V Liner 8EEE
19-167-445 Leather, Dupont Kevlar Aramid Fiber, CrossTech 9.5EEE
19-167-444 Pro-V Liner 9EEE
19-167-449 Leather, Dupont Kevlar Aramid Fiber, CrossTech 11.5EEE
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 19 options available.
19 options
Catalog No. 19-167-455 Supplier Honeywell™ Supplier No. BT5006B1605
Add to Cart
Add to Cart
Superior combination of comfort and absorbency
Quilted to 7.5 oz Kevlar™ felt thermal barrier and laminated to fully encapsulating Crosstech™ Footwear Fabric bootie. Crosstech™ moisture barrier provides superior liquid penetration resistance to blood, body fluids and NFPA tested common chemicals.
  • Military AB leather construction insulates and adds abrasion-, puncture-, water- and chemical-resistance
  • With heavy duty 0.75 in. reinforced straps mounted to the exterior of boot and anchored with a 2 × 1.5 in. shield-shaped base
  • External texon board shinguard protects and guards sensitive shin area of legs
  • Patented PowerToe™ cover resists toe abrasion and provides unparalled wear over the lifetime of the boots
  • Oblique toe safety cap provides additional room in height and width of boot
  • Fire/Ice PowerSole™ angled lugs provide substantial edging and gripping capabilities
  • Molded thermoplastic heel counter locks foot into the boot and anchors it in place
  • Patented PowerHeel™ cover protects heel area from abusive doffing of boots
  • Corrison-resistant steel ladder shank and stainless steel, full length, toe-to-heel puncture resistant bottom plate
Order Info
D and E widths also available.
Compliance
Meet NFPA 1971, 2007 edition requirements
TRUSTED_SUSTAINABILITY
Specifications
Height (English) 14 in.
Material Leather, Dupont Kevlar Aramid Fiber, CrossTech
Size 16EEE