Learn More
Honeywell™ PRO 5007 Structural Firefighting Bunker Boots - C Width
Catalog No. 18821415
Change view
Click to view available options
Size:
10.5C
10C
11.5C
11C
12C
13C
14C
6.5C
6C
7.5C
7C
8.5C
8C
9.5C
9C
15 product options available for selection
Product selection table with 15 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
18-821-415 14C
18-821-407 9C
18-821-403 7C
18-821-405 8C
18-821-406 8.5C
18-821-402 6.5C
18-821-401 6C
18-821-404 7.5C
18-821-408 9.5C
18-821-409 10C
18-821-410 10.5C
18-821-411 11C
18-821-413 12C
18-821-412 11.5C
18-821-414 13C
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 15 options available.
15 options
Catalog No. 18-821-415 Supplier Honeywell™ Supplier No. BT50071401
Add to Cart
Add to Cart
Superior combination of comfort and absorbency
  • Quilted to 7.5 oz. Kevlar™ felt thermal barrier and laminated to fully encapsulating Crosstech™ Footwear Fabric bootie
  • Crosstech™ moisture barrier provides superior liquid penetration resistance to blood, body fluids and NFPA tested common chemicals
  • Military AB leather construction insulates and adds abrasion-, puncture-, water- and chemical-resistance
  • With heavy duty 0.75 in. reinforced straps mounted to the exterior of boot and anchored with a 2 × 1.5 in. shield-shaped base
  • External texon board shinguard protects and guards sensitive shin area of legs
  • Patented PowerToe™ cover resists toe abrasion and provides unparalled wear over the lifetime of the boots
  • Oblique toe safety cap provides additional room in height and width of boot
  • Fire/Ice PowerSole™ angled lugs provide substantial edging and gripping capabilities
  • Molded thermoplastic heel counter locks foot into the boot and anchors it in place
  • Patented PowerHeel™ cover protects heel area from abusive doffing of boots
  • Corrison-resistant steel ladder shank and stainless steel, full length, toe-to-heel puncture resistant bottom plate
Compliance
Meet NFPA 1971, 2007 edition requirements
TRUSTED_SUSTAINABILITY
Specifications
Height (English) 14 in.
Size 14C