Learn More
Honeywell™ PRO 5007 Structural Firefighting Bunker Boots - D Width
Catalog No. 18821416
Change view
Click to view available options
Size:
10.5D
10D
11.5D
11D
12.5D
12D
13D
14D
15D
4.5DC
4D
5.5D
5D
6.5D
6DC
7.5D
7D
8.5D
8D
9.5D
9D
21 product options available for selection
Product selection table with 21 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
18-821-416 4D
18-821-417 4.5DC
18-821-418 5D
18-821-422 7D
18-821-420 6DC
18-821-421 6.5D
18-821-423 7.5D
18-821-419 5.5D
18-821-424 8D
18-821-425 8.5D
18-821-426 9D
18-821-427 9.5D
18-821-428 10D
18-821-431 11.5D
18-821-429 10.5D
18-821-430 11D
18-821-432 12D
18-821-434 13D
18-821-433 12.5D
18-821-435 14D
18-821-436 15D
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 21 options available.
21 options
Catalog No. 18-821-416 Supplier Honeywell™ Supplier No. BT50070402
Add to Cart
Add to Cart
Quilted to 7.5 oz. Kevlar™ felt thermal barrier and laminated to fully encapsulating Crosstech™ Footwear Fabric bootie
  • Crosstech™ moisture barrier provides superior liquid penetration resistance to blood, body fluids and NFPA tested common chemicals
  • Military AB leather construction insulates and adds abrasion-, puncture-, water- and chemical-resistance
  • With heavy duty 0.75 in. reinforced straps mounted to the exterior of boot and anchored with a 2 × 1.5 in. shield-shaped base
  • External texon board shinguard protects and guards sensitive shin area of legs
  • Patented PowerToe™ cover resists toe abrasion and provides unparalled wear over the lifetime of the boots
  • Oblique toe safety cap provides additional room in height and width of boot
  • Fire/Ice PowerSole™ angled lugs provide substantial edging and gripping capabilities
  • Molded thermoplastic heel counter locks foot into the boot and anchors it in place
  • Patented PowerHeel™ cover protects heel area from abusive doffing of boots
  • Corrison-resistant steel ladder shank and stainless steel, full length, toe-to-heel puncture resistant bottom plate
Compliance
Meet NFPA 1971, 2007 edition requirements
TRUSTED_SUSTAINABILITY
Specifications
Height (English) 14 in.
Size 4D