Learn More
Honeywell™ PRO 5050 Struximity Leather Boots, Berry Compliant
Catalog No. 19813901
Change view
Click to view available options
Size:
10.5C
10.5D
10C
10D
11.5C
11.5D
11C
12.5C
12.5D
12C
13C
13D
14C
14D
15D
16D
5.5D
5D
6.5D
6D
7.5D
7D
8.5D
8D
9.5C
9.5D
9C
9D
28 product options available for selection
Product selection table with 28 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-813-901 5D
19-813-902 5.5D
19-813-903 6D
19-813-904 6.5D
19-813-905 7D
19-813-906 7.5D
19-813-907 8D
19-813-910 9.5D
19-813-908 8.5D
19-813-909 9D
19-813-911 10D
19-813-912 10.5D
19-813-914 11.5D
19-813-916 12.5D
19-813-917 13D
19-813-919 15D
19-813-920 16D
19-813-963 10C
19-813-962 9.5C
19-813-961 9C
19-813-964 10.5C
19-813-965 11C
19-813-966 11.5C
19-813-967 12C
19-813-968 12.5C
19-813-969 13C
19-813-970 14C
19-813-918 14D
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 28 options available.
28 options
Catalog No. 19-813-901 Supplier Honeywell™ Supplier No. BT5050B0502
Add to Cart
Add to Cart
Lightweight, comfortable, high-speed, leather pull-on bunker boots dual certified for both Structural and Proximity (ARFF) firefighting
Made of high performance fire resistant and waterproof leather and Goodyear welt made to withstand the harsh proximity environment
  • Athletic construction delivers unencumbered performance and agility
  • Full height bootie combines an aluminized radiant shield, 7.5 oz Kevlar thermal felt quilted to Cambrelle™ liner and laminated to Crosstech™ Moisture barrier fabric for protection from heat, fluids, and internal abrasion
  • Specially designed profiled boot shaft is large enough to accommodate large calves while still fitting comfortably under proximity pants
  • Vibram™ lug replaceable soles with Fire and Ice compound are cemented and security stitched using Goodyear welt construction for superior durability protection
  • Fire and Ice sole compound improves winter traction on ice and snow-even on wet ice
  • Steel bottom plate and shank offer protection from puncture and dangerous twisting
  • Improved heel pocket and comfortable molded insert defend against heel slippage
  • APT Barcode — Departmental NFPA 1851 Tracking
  • Certified to NFPA 1971, current edition for Proximity and Structural Fire Fighting
  • Weight: 6.6 pounds
  • Union made in USA
  • All components are Berry Amendment compliant
TRUSTED_SUSTAINABILITY
Specifications
Liner Material Cambrelle
Material Leather, CrossTech, Dupont Kevlar Aramid Fiber
Size 5D