Learn More
Honeywell™ PRO 5050 Struximity Leather Boots, Berry-Compliant, Wide
Catalog No. 19813960
Change view
Click to view available options
Material:
Leather, CrossTech, Dupont Kevlar Aramid Fiber
Leather, Dupont Kevlar Aramid Fiber, CrossTech
Size:
10.5E
10.5EEE
10E
10EEE
11.5E
11.5EEE
11E
11EEE
12.5E
12.5EEE
12E
12EEE
13E
13EEE
14E
14EEE
15E
15EEE
16E
16EEE
5.5E
5.5EEE
5E
5EEE
6.5E
6.5EEE
6E
6EEE
7.5E
7.5EEE
7E
7EEE
8.5E
8.5EEE
8E
8EEE
9.5E
9.5EEE
9E
9EEE
40 product options available for selection
Product selection table with 40 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material Size
19-813-960 Leather, CrossTech, Dupont Kevlar Aramid Fiber 16EEE
19-813-921 Leather, Dupont Kevlar Aramid Fiber, CrossTech 5E
19-813-922 Leather, CrossTech, Dupont Kevlar Aramid Fiber 5.5E
19-813-923 Leather, CrossTech, Dupont Kevlar Aramid Fiber 6E
19-813-925 Leather, CrossTech, Dupont Kevlar Aramid Fiber 7E
19-813-924 Leather, CrossTech, Dupont Kevlar Aramid Fiber 6.5E
19-813-926 Leather, CrossTech, Dupont Kevlar Aramid Fiber 7.5E
19-813-927 Leather, CrossTech, Dupont Kevlar Aramid Fiber 8E
19-813-928 Leather, CrossTech, Dupont Kevlar Aramid Fiber 8.5E
19-813-930 Leather, CrossTech, Dupont Kevlar Aramid Fiber 9.5E
19-813-929 Leather, CrossTech, Dupont Kevlar Aramid Fiber 9E
19-813-931 Leather, CrossTech, Dupont Kevlar Aramid Fiber 10E
19-813-934 Leather, CrossTech, Dupont Kevlar Aramid Fiber 11.5E
19-813-933 Leather, CrossTech, Dupont Kevlar Aramid Fiber 11E
19-813-932 Leather, CrossTech, Dupont Kevlar Aramid Fiber 10.5E
19-813-935 Leather, CrossTech, Dupont Kevlar Aramid Fiber 12E
19-813-936 Leather, CrossTech, Dupont Kevlar Aramid Fiber 12.5E
19-813-938 Leather, CrossTech, Dupont Kevlar Aramid Fiber 14E
19-813-937 Leather, CrossTech, Dupont Kevlar Aramid Fiber 13E
19-813-939 Leather, CrossTech, Dupont Kevlar Aramid Fiber 15E
19-813-940 Leather, CrossTech, Dupont Kevlar Aramid Fiber 16E
19-813-941 Leather, CrossTech, Dupont Kevlar Aramid Fiber 5EEE
19-813-942 Leather, CrossTech, Dupont Kevlar Aramid Fiber 5.5EEE
19-813-943 Leather, CrossTech, Dupont Kevlar Aramid Fiber 6EEE
19-813-944 Leather, CrossTech, Dupont Kevlar Aramid Fiber 6.5EEE
19-813-945 Leather, CrossTech, Dupont Kevlar Aramid Fiber 7EEE
19-813-946 Leather, CrossTech, Dupont Kevlar Aramid Fiber 7.5EEE
19-813-949 Leather, CrossTech, Dupont Kevlar Aramid Fiber 9EEE
19-813-947 Leather, CrossTech, Dupont Kevlar Aramid Fiber 8EEE
19-813-948 Leather, CrossTech, Dupont Kevlar Aramid Fiber 8.5EEE
19-813-950 Leather, CrossTech, Dupont Kevlar Aramid Fiber 9.5EEE
19-813-951 Leather, CrossTech, Dupont Kevlar Aramid Fiber 10EEE
19-813-952 Leather, CrossTech, Dupont Kevlar Aramid Fiber 10.5EEE
19-813-954 Leather, CrossTech, Dupont Kevlar Aramid Fiber 11.5EEE
19-813-953 Leather, CrossTech, Dupont Kevlar Aramid Fiber 11EEE
19-813-955 Leather, CrossTech, Dupont Kevlar Aramid Fiber 12EEE
19-813-958 Leather, CrossTech, Dupont Kevlar Aramid Fiber 14EEE
19-813-957 Leather, CrossTech, Dupont Kevlar Aramid Fiber 13EEE
19-813-956 Leather, CrossTech, Dupont Kevlar Aramid Fiber 12.5EEE
19-813-959 Leather, CrossTech, Dupont Kevlar Aramid Fiber 15EEE
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 40 options available.
40 options
Catalog No. 19-813-960 Supplier Honeywell™ Supplier No. BT5050B1605
Add to Cart
Add to Cart
Lightweight, comfortable, high-speed, wide-width leather pull-on bunker boots dual certified for both Structural and Proximity (ARFF) firefighting
Made of high performance fire resistant and waterproof leather and Goodyear welt made to withstand the harsh proximity environment
  • Athletic construction delivers unencumbered performance and agility
  • Full height bootie combines an aluminized radiant shield, 7.5 oz Kevlar thermal felt quilted to Cambrelle™ liner and laminated to Crosstech™ Moisture barrier fabric for protection from heat, fluids, and internal abrasion
  • Specially designed profiled boot shaft is large enough to accommodate large calves while still fitting comfortably under proximity pants
  • Vibram™ lug replaceable soles with Fire and Ice compound are cemented and security stitched using Goodyear welt construction for superior durability protection
  • Fire and Ice sole compound improves winter traction on ice and snow
  • Steel bottom plate and shank offer protection from puncture and dangerous twisting
  • Improved heel pocket and comfortable molded insert defend against heel slippage
  • APT Barcode — Departmental NFPA 1851 Tracking
  • Certified to NFPA 1971, current edition for Proximity and Structural Fire Fighting
  • Weight: 6.6 pounds
  • Wide width
  • Union made in USA. All components are Berry Amendment compliant
TRUSTED_SUSTAINABILITY
Specifications
Liner Material Cambrelle
Material Leather, CrossTech, Dupont Kevlar Aramid Fiber
Size 16EEE
Weight (English) 6.6 lb.