Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Honeywell™ Pure Fit™ 380 Nitrile Coated Nylon Gloves
Description
- Constructed of lightweight, seamless knit nylon
- Dipped palm and fingertips improve dexterity, and reduce hand fatigue
- Coating provides an extra wear area for extended use, and increased comfort
- Low linting for cleanroom applications
Specifications
Specifications
| Coated Area | Palm, Fingers |
| Construction | Seamless Knit |
| Coating Material | Nitrile |
| Color | Black |
| Exterior Finish | No Texture |
| Hand | Specific |
| Material | Nylon |
| Product Type | Seamless Knit Gloves |
| Size | X-Large |
| For Use With (Application) | Parts Assembly/Handling, Automotive Assembly, Painting, and Power Tool Operation |
| Show More |
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?