Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell™ Pure Fit™ Nitrile-Coated Dyneema Gloves

Catalog No. p-145760
Change view
Click to view available options
Size:
Small
Glove Cuff Color:
Red
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Glove Cuff Color
19-822-658 Small Red
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-822-658 Supplier Honeywell™ Supplier No. PF545S
This item is not returnable. View return policy

Lightweight, breathable fibers provide tactile sensitivity for precision handling and delicate operations.

  • Dyneema fiber provides cut and abrasion resistance in light-duty applications
  • Blue nitrile coating provides grip plus additional resistance to cuts and abrasions
  • Machine washable for longer service life

TRUSTED_SUSTAINABILITY

Specifications

Color Blue, White
Cut Resistance Rating ANSI A2
Material HPPE
Product Type Cut-resistant Gloves
Size Small
EN388 Standard EN 388:4242
For Use With (Application) construction, recycling, lumber and mill work, Glass handling, Metal stamping and automotive assembly
Glove Cuff Color Red
Model PF545-S
Series Pure Fit™
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.