Learn More
Honeywell™ Ranger™ 7022 and 5128 Structural Pull On Bunker Boots
Catalog No. 19814651
Change view
Click to view available options
Steel Safe-Toe; 10
Catalog No.
19-814-651 Steel Safe-Toe; 10
1 options
Catalog No. 19-814-651 Supplier Honeywell™ Supplier No. BT512810
Combat L.B.™ (Light Bunker) with Shoe-Fit™ construction
  • Fire-grade rubber
  • Nomex™/ Aramid™ side panels
  • Cambrelle™ lining
  • Duratech™ membrane barrier
  • Kevlar™/ Nomex Foot Liner
  • Urethane/ Silicone Enhanced Insulation
  • Reinforced Leather Pull-Up Loops
  • Removable Cushion Insoles
  • Compatible Patented DRYZ™ Insert (Optional)
  • Built-in Shin Protector
  • Foot Guard Protection in Metatarsal Area
  • Wide Angle Lug Outsole
  • Steel Safe-Toe™
  • Steel Protective Midsole
  • Triple Ribbed Steel Shank
  • Barcoded for easy tracking
  • Meets ASTM F2413-05; UL Listed; Made in USA
  • Certifications: NFPA 1971, 2007 Edition, for Structural Fire Fighting; Optional NFPA 1992, 2005 Edition, for Liquid Splash/ HazMat Emergencies (must be requested at time of purchase)
TRUSTED_SUSTAINABILITY
Specifications
Height (English) 16 in.
Material Dupont Kevlar Aramid Fiber, Dupont Nomex Aramid Fiber Blend, Wool
Size 10