Learn More
MSC Honeywell Safety Glasses, 50% Gray Anti-Fog Lens, Black Frame, HydroShield AF
Catalog No. 5000508485
Encompass
Uvex Genesis safety glasses are engineered for comfort, protection, and style, making them a top-selling premium eyewear. Built with Multi-Material Technology (MMT) for comfort and protection, they offer adjustable features and a wide range of lens tints for extraordinary all-day comfort and compliance. Made in the USA.
  • Easy, economical lens replacement system
  • Lifetime frame guarantee
  • Sleek ¾ frame for high performance
  • Meets ANSI Z87.1+ - 2015 and CSA Z94.3 2015 standards
  • HydroShield anti-fog coating offers 90X longer fog-free time and 2X greater scratch resistance
  • Dura-streme dual coating for 3X longer lens life and 2X greater scratch resistance
  • Soft browbar, flexible nose bridge, and adjustable temple length
Catalog No. 50-005-08485 Supplier MSC Supplier No. 32474603
Uvex Genesis safety glasses are engineered for comfort, protection, and style, making them a top-selling premium eyewear. Built with Multi-Material Technology (MMT) for comfort and protection, they offer adjustable features and a wide range of lens tints for extraordinary all-day comfort and compliance. Made in the USA.
  • Easy, economical lens replacement system
  • Lifetime frame guarantee
  • Sleek ¾ frame for high performance
  • Meets ANSI Z87.1+ - 2015 and CSA Z94.3 2015 standards
  • HydroShield anti-fog coating offers 90X longer fog-free time and 2X greater scratch resistance
  • Dura-streme dual coating for 3X longer lens life and 2X greater scratch resistance
  • Soft browbar, flexible nose bridge, and adjustable temple length