Learn More
MSC Honeywell Safety Glasses, Genesis, 50% Gray, Anti-Fog, Black Frame
Catalog No. 5000508483
Encompass
Uvex Genesis Safety Glasses offer comfort, protection, and style. Built with Multi-Material Technology and a wide assortment of lens tints, these glasses provide all-day comfort and high compliance, Made in the USA.
  • Delivers superior all-day comfort, protection, and style.
  • Features an easy and economical lens replacement system.
  • Frames include a lifetime guarantee and ¾ frame design.
  • Meets ANSI Z87.1+ - 2015 and CSA Z94.3 2015 standards.
  • Polycarbonate lens with various coating options like Hydroshield AF, Dura-streme AF/HC, Uvextreme.
  • Includes ratchet, adjustable temples, flexible nose, and Multi-Material Technology.
  • Available in a wide range of lens tints and frame colors.
Catalog No. 50-005-08483 Supplier MSC Supplier No. 32393845
Uvex Genesis Safety Glasses offer comfort, protection, and style. Built with Multi-Material Technology and a wide assortment of lens tints, these glasses provide all-day comfort and high compliance, Made in the USA.
  • Delivers superior all-day comfort, protection, and style.
  • Features an easy and economical lens replacement system.
  • Frames include a lifetime guarantee and ¾ frame design.
  • Meets ANSI Z87.1+ - 2015 and CSA Z94.3 2015 standards.
  • Polycarbonate lens with various coating options like Hydroshield AF, Dura-streme AF/HC, Uvextreme.
  • Includes ratchet, adjustable temples, flexible nose, and Multi-Material Technology.
  • Available in a wide range of lens tints and frame colors.