Learn More
MSC Honeywell Safety Replacement Lens, HydroShield Anti-Fog, Amber, Polycarbonate
Catalog No. 5000508535
Encompass
This replacement lens, designed for Genesis Protective Eyewear, features HydroShield Anti-Fog coating and is made from durable amber polycarbonate. It offers universal sizing for broad compatibility and provides clear vision in various working conditions.
  • HydroShield anti-fog lens coating
  • Amber lens for enhanced contrast
  • Polycarbonate material for durability
  • Universal size for versatile use
  • Designed for Genesis protective eyewear
Catalog No. 50-005-08535 Supplier MSC Supplier No. 33817511
This replacement lens, designed for Genesis Protective Eyewear, features HydroShield Anti-Fog coating and is made from durable amber polycarbonate. It offers universal sizing for broad compatibility and provides clear vision in various working conditions.
  • HydroShield anti-fog lens coating
  • Amber lens for enhanced contrast
  • Polycarbonate material for durability
  • Universal size for versatile use
  • Designed for Genesis protective eyewear