Learn More
Honeywell™ Swat Tactical Gloves
Catalog No. 191600144
Change view
Click to view available options
Size:
7
8
10
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-160-0144 7
19-160-0145 8
19-160-0147 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 19-160-0144 Supplier Honeywell™ Supplier No. 16407
This item is not returnable. View return policy
Gloves are constructed of DuPont™ Nomex™ for flame and fire resistance, with Aramid Fiber stitching for additional strength.
  • DuPont™ Nomex™ construction, sewn with DuPont™ Kevlar™ Aramid Fiber
  • Chrome-tanned sheepskin leather palm for enhanced grip capabilities
TRUSTED_SUSTAINABILITY
Specifications
Color Black
Material Nomex
Size 7
Gender Unisex
Glove Cuff Style Gauntlet
Palm Type Sheepskin Leather
Series 1640