Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell™ Tails Tan Nomex BPR Turnout Gear

Catalog No. 19300139
Change view
Click to view available options
Pants; Nomex; Tan; BPR 2402
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
19-300-139 Pants; Nomex; Tan; BPR 2402
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-300-139 Supplier Honeywell™ Supplier No. SAEGSA00566

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Lighter weight with same protection, greater mobility, and reduced stress load

  • 7.5 ox. Tan Nomex outer shell
  • Aralite/Nomes thermal liner
  • Crosstech™/Nomex™ moisture barrier
  • Inspection port liner
  • Detachable liner
  • Two take-up postman straps
Coat
  • Foldover comfort chinstrap
  • Cuffs
  • 1.5 in. interior hooks and D's Velcro clsoure
  • Pocket label liner
  • Nomex™-tabbed long wristlets
  • Articulating rapid rescue strap
  • Lime Scotchlite™ trim
  • Handwarmer pockets
  • 8 x 3 x 2 in. radio pocket on left chest

Pants
  • Narrow 1.5 in. Velcro fly
  • 2 in. lime Scotchlite™ cuff trim
  • 9 x9 x 1.5 in. bellows pockets

Order Info

Please specify sizes when ordering: small, medium, large, XL, XXL, XXXL, XXXXL

Compliance

NFPA 1971, 2007 Edition

TRUSTED_SUSTAINABILITY
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.