The Honeywell Uvex 880 Series Klip Lifts are designed to clip easily onto hard hat or bump cap brims and are worn with existing protective eyewear. This full-front, cobalt blue, shade 6 lens offers quick and convenient vision enhancement and protection, making it suitable for industrial applications such as steelmaking, welding, cutting, foundries, and glassmaking operations.
- Clips easily onto hard hat or bump cap brims.
- Worn with existing protective eyewear.
- Flip-up design allows quick vision enhancement or removal from line of sight.
- Offers a full 180° arc of movement.
- Integral spring clip holds the lens securely without tools.
- Cobalt blue lens provides enhanced visibility and glare protection.
- Shade 6 provides appropriate shading for welding tasks.
- Durable stainless steel construction.
- Lenses offer 99.9% ultra-violet protection.