Learn More
MSC Honeywell Uvex Safety Glasses, Clear Lens, Black Frame, Anti-Fog, Sealed
Catalog No. 5000508482
Encompass
These anti-fog sealed safety glasses are designed for high-particulate environments, offering protection against dust, dirt, and debris. They feature an airtight, sealed construction with a removable foam frame for a snug and secure fit, along with indirect ventilation channels for superior dust ventilation. The glasses are impact-resistant and protect against wind, debris, and glare.
  • Air-tight, sealed fit with removable foam frame.
  • Protects against impact, wind, debris, and glare.
  • Features anti-fog hard coating.
  • Dielectric frame provides electrical insulation.
  • Meets CSA Z94.3 and ANSI Z.87.1 standards.
  • Offers 99.9% UV protection (U6 ranked).
  • Indirect ventilation channels on the subframe provide superior dust ventilation.
  • Ideal for use in agriculture, construction, manufacturing, and mining.
Catalog No. 50-005-08482 Supplier MSC Supplier No. 32033755
These anti-fog sealed safety glasses are designed for high-particulate environments, offering protection against dust, dirt, and debris. They feature an airtight, sealed construction with a removable foam frame for a snug and secure fit, along with indirect ventilation channels for superior dust ventilation. The glasses are impact-resistant and protect against wind, debris, and glare.
  • Air-tight, sealed fit with removable foam frame.
  • Protects against impact, wind, debris, and glare.
  • Features anti-fog hard coating.
  • Dielectric frame provides electrical insulation.
  • Meets CSA Z94.3 and ANSI Z.87.1 standards.
  • Offers 99.9% UV protection (U6 ranked).
  • Indirect ventilation channels on the subframe provide superior dust ventilation.
  • Ideal for use in agriculture, construction, manufacturing, and mining.