Learn More
MSC Honeywell VeriShield Earmuffs, Helmet Capmount, NRR 24, Black
Catalog No. 5000509112
Encompass
These passive earmuffs provide essential hearing protection, designed for optimum comfort during extended use. They feature a wide cup opening with memory foam ear cushions and a lightweight cushioned headband, ensuring a comfortable fit for a wide range of head sizes.
  • Lightweight cushioned headband features steel-wire construction and precision micro-adjustments for a proper fit.
  • Memory foam ear cushions combine denser and softer foam for greater comfort and attenuation, reducing squeezing pressure.
  • Robust construction includes an oil-resistant cup surface and ABS molded design for lighter weight.
  • Air Flow Control technology provides optimum attenuation across all frequencies.
  • Noise reduction rating (NRR) of 24.
Catalog No. 50-005-09112 Supplier MSC Supplier No. 95460473
These passive earmuffs provide essential hearing protection, designed for optimum comfort during extended use. They feature a wide cup opening with memory foam ear cushions and a lightweight cushioned headband, ensuring a comfortable fit for a wide range of head sizes.
  • Lightweight cushioned headband features steel-wire construction and precision micro-adjustments for a proper fit.
  • Memory foam ear cushions combine denser and softer foam for greater comfort and attenuation, reducing squeezing pressure.
  • Robust construction includes an oil-resistant cup surface and ABS molded design for lighter weight.
  • Air Flow Control technology provides optimum attenuation across all frequencies.
  • Noise reduction rating (NRR) of 24.