Learn More
MSC Honeywell Water Additive, 8 oz Bottle, Preserved Solution, 30-Month Shelf Life
Catalog No. 5000507893
Encompass
This replacement water additive is designed for use with Porta Stream I, II, and III eyewash stations. It is formulated to be mixed with clean, potable water, creating a preserved solution for emergency eye rinsing. Each 8 oz bottle can treat 14-16 gallons of water, ensuring compliance with ANSI Z358.1 requirements for a full 15-minute flushing period when used with Porta Stream II and III.
  • Mixes with clean, potable water to create a preserved solution.
  • Does not require costly plumbing for installation.
  • Provides constant fluid flow and stream height for 15-minute flushing.
  • One 8 oz bottle treats 14-16 gallons of water.
  • Unopened shelf life of 30 months; 6 months once opened.
  • For use with compatible eyewash stations.
  • Recommended mixing ratio is 10:1 (additive to water).
  • Station cleaning required before refilling.
Catalog No. 50-005-07893 Supplier MSC Supplier No. 02986735
This replacement water additive is designed for use with Porta Stream I, II, and III eyewash stations. It is formulated to be mixed with clean, potable water, creating a preserved solution for emergency eye rinsing. Each 8 oz bottle can treat 14-16 gallons of water, ensuring compliance with ANSI Z358.1 requirements for a full 15-minute flushing period when used with Porta Stream II and III.
  • Mixes with clean, potable water to create a preserved solution.
  • Does not require costly plumbing for installation.
  • Provides constant fluid flow and stream height for 15-minute flushing.
  • One 8 oz bottle treats 14-16 gallons of water.
  • Unopened shelf life of 30 months; 6 months once opened.
  • For use with compatible eyewash stations.
  • Recommended mixing ratio is 10:1 (additive to water).
  • Station cleaning required before refilling.