Learn More
MSC Honeywell Work Gloves, 15 Gauge, Nylon, Palm Foamed PVC Coating, Medium
Catalog No. 5000507340
Encompass
The NorthFlex Red NF11 is an ultra-lightweight, general-purpose glove with a 15 gauge seamless nylon shell and foamed PVC coating. The palm-dipped PVC provides soft, flexible comfort and abrasion resistance. Its unique foamed PVC texture offers superior grip in wet conditions, enhancing dexterity and tactility.
  • Foamed PVC coating provides an exceptional combination of abrasion resistance and grip, especially in wet conditions.
  • Stretchy and form-fitting shell for all-day comfort.
  • Bright red color for greater visibility.
  • Polyester shell material helps wick away moisture and provides superior comfort.
  • PVC coating offers high tactile sensitivity and excellent grip in dry and wet applications.
Catalog No. 50-005-07340 Supplier MSC Supplier No. 79195848
The NorthFlex Red NF11 is an ultra-lightweight, general-purpose glove with a 15 gauge seamless nylon shell and foamed PVC coating. The palm-dipped PVC provides soft, flexible comfort and abrasion resistance. Its unique foamed PVC texture offers superior grip in wet conditions, enhancing dexterity and tactility.
  • Foamed PVC coating provides an exceptional combination of abrasion resistance and grip, especially in wet conditions.
  • Stretchy and form-fitting shell for all-day comfort.
  • Bright red color for greater visibility.
  • Polyester shell material helps wick away moisture and provides superior comfort.
  • PVC coating offers high tactile sensitivity and excellent grip in dry and wet applications.