Learn More
MSC Honeywell Work Gloves, Nylon, Nitrile Coated, Large
Catalog No. 5000507014
Encompass
Honeywell Pure Fit 13 Nylon Black Nitrile Palm & Fingertips Gray 375 gloves are general purpose gloves designed for precision work. They feature a lightweight, low-linting nylon liner and a nitrile palm and fingertip coating, offering excellent dexterity, tactile sensitivity, and abrasion resistance. The comfortable and breathable design helps reduce hand fatigue.
  • Low-linting, lightweight nylon seamless knit liners.
  • High tensile strength nylon provides abrasion resistance.
  • Nitrile palm and fingertip coating for enhanced grip and protection.
  • Palm-dipped design reduces hand fatigue and offers extreme dexterity.
  • Provides ultimate dexterity and tactile sensitivity for precision work.
  • Lightweight and comfortable, feeling like a second skin.
  • Coating creates an extra wear area for longer wear.
  • Knit wrist cuff for a secure fit.
Catalog No. 50-005-07014 Supplier MSC Supplier No. 72916505
Honeywell Pure Fit 13 Nylon Black Nitrile Palm & Fingertips Gray 375 gloves are general purpose gloves designed for precision work. They feature a lightweight, low-linting nylon liner and a nitrile palm and fingertip coating, offering excellent dexterity, tactile sensitivity, and abrasion resistance. The comfortable and breathable design helps reduce hand fatigue.
  • Low-linting, lightweight nylon seamless knit liners.
  • High tensile strength nylon provides abrasion resistance.
  • Nitrile palm and fingertip coating for enhanced grip and protection.
  • Palm-dipped design reduces hand fatigue and offers extreme dexterity.
  • Provides ultimate dexterity and tactile sensitivity for precision work.
  • Lightweight and comfortable, feeling like a second skin.
  • Coating creates an extra wear area for longer wear.
  • Knit wrist cuff for a secure fit.