Learn More
Medchemexpress LLC HY-N0003 100mg , Honokiol CAS:35354-74-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN0003100MG
Encompass_Preferred
HY-N0003 100mg Honokiol CAS: 35354-74-6 Honokiol is a bioactive, biphenolic phytochemical that possesses potent antioxidative, anti-inflammatory, antiangiogenic, and anticancer activities by targeting a variety of signaling molecules. It inhibits the activation of Akt. Honokiol can readily cross the blood brain barrier[4]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1164