Learn More
Apexbio Technology LLC Honokiol 35354-74-6 20mg
SDP
Catalog No. 502240959
Encompass_Preferred
Honokiol (CAS 35354-74-6) is a small-molecule inhibitor targeting nuclear factor-kappa B (NF- B) It is designed to modulate NF- B activation induced by stimuli such as TNF and okadaic acid thereby regulating inflammatory and oncogenic signaling pathways Honokiol exerts its biological activity primarily through inhibition of NF- B activation and free radical scavenging In cell-based studies honokiol demonstrates antiproliferative and antiangiogenic activity with IC50 values ranging from approximately 10 M to 50 M depending on cell types and experimental conditions Based on these pharmacological properties honokiol holds research potential in studying inflammation signaling oxidative stress mechanisms tumor biology and angiogenesis inhibition
This item is not returnable. View return policy
Catalog No. 50-224-0959 Supplier Apexbio Technology LLC Supplier No. N167220
Honokiol (CAS 35354-74-6) is a small-molecule inhibitor targeting nuclear factor-kappa B (NF- B) It is designed to modulate NF- B activation induced by stimuli such as TNF and okadaic acid thereby regulating inflammatory and oncogenic signaling pathways Honokiol exerts its biological activity primarily through inhibition of NF- B activation and free radical scavenging In cell-based studies honokiol demonstrates antiproliferative and antiangiogenic activity with IC50 values ranging from approximately 10 M to 50 M depending on cell types and experimental conditions Based on these pharmacological properties honokiol holds research potential in studying inflammation signaling oxidative stress mechanisms tumor biology and angiogenesis inhibition