Learn More
Medchemexpress LLC Honokiol dichloroacetate | 1620160-42-0 | 99.1% | 488.19 g/mol | C22H18Cl4O4 | 100MG
SDP
Catalog No. 5000235736
Encompass_Preferred
Honokiol dichloroacetate is a bis-dichloroacetate ester analog of honokiol used as a small-molecule research reagent. It has demonstrated in vitro activity against human prostate cancer cells and is used as a biochemical tool compound to study mitochondrial and androgen receptor-related pathways.
  • Dichloroacetate ester of honokiol.
  • Reported to inhibit prostate cancer cell growth in vitro.
  • Reduces androgen receptor (AR) protein levels in cell studies.
  • High purity (99.1%) determined by RP-HPLC.
  • Molecular weight 488.19 g/mol; formula C22H18Cl4O4.
  • Available in multiple pack sizes, including 100 mg.
  • Intended for research use only.
Catalog No. 50-002-35736 Supplier Medchemexpress LLC Supplier No. HY124292100MG
Honokiol dichloroacetate is a bis-dichloroacetate ester analog of honokiol used as a small-molecule research reagent. It has demonstrated in vitro activity against human prostate cancer cells and is used as a biochemical tool compound to study mitochondrial and androgen receptor-related pathways.
  • Dichloroacetate ester of honokiol.
  • Reported to inhibit prostate cancer cell growth in vitro.
  • Reduces androgen receptor (AR) protein levels in cell studies.
  • High purity (99.1%) determined by RP-HPLC.
  • Molecular weight 488.19 g/mol; formula C22H18Cl4O4.
  • Available in multiple pack sizes, including 100 mg.
  • Intended for research use only.