Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Honokiol dichloroacetate | 1620160-42-0 | 99.1% | 488.19 g/mol | C22H18Cl4O4 | 50MG
SDP

Catalog No. 5000237038
Encompass_Preferred

Honokiol DCA (honokiol dichloroacetate) is a dichloroacetate ester of honokiol used as a research reagent with reported in vitro activity against prostate cancer cell growth and androgen receptor expression. The compound is provided as a well-characterized, high-purity material suitable for laboratory studies.

  • Research-grade dichloroacetate ester of honokiol.
  • CAS 1620160-42-0; molecular weight 488.19 g/mol.
  • Chemical formula C22H18Cl4O4.
  • Purity 99.1% (manufacturer reported).
  • Soluble in DMSO (100 mg/mL); in vivo formulation protocols available.
  • Storage: -20°C protected from light; in solvent -80°C (6 months) or -20°C (1 month).
  • Reported to inhibit prostate cancer cell growth and reduce androgen receptor levels in vitro.

Catalog No. 50-002-37038 Supplier Medchemexpress LLC Supplier No. HY12429250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Honokiol DCA (honokiol dichloroacetate) is a dichloroacetate ester of honokiol used as a research reagent with reported in vitro activity against prostate cancer cell growth and androgen receptor expression. The compound is provided as a well-characterized, high-purity material suitable for laboratory studies.

  • Research-grade dichloroacetate ester of honokiol.
  • CAS 1620160-42-0; molecular weight 488.19 g/mol.
  • Chemical formula C22H18Cl4O4.
  • Purity 99.1% (manufacturer reported).
  • Soluble in DMSO (100 mg/mL); in vivo formulation protocols available.
  • Storage: -20°C protected from light; in solvent -80°C (6 months) or -20°C (1 month).
  • Reported to inhibit prostate cancer cell growth and reduce androgen receptor levels in vitro.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.