Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honokiol, Tocris Bioscience™

Catalog No. 459010
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45-901-0 10 mg
45-905-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 45-901-0 Supplier R&D Systems Supplier No. 4590/10

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This item is not returnable. View return policy

Blocks NF-κB activation; also anti-inflammatory and antioxidant

Anti-inflammatory, antioxidant, antitumor and antiangiogenic agent. Blocks NF-κB activation by a variety of stimuli, including TNF and okadaic acid. Shown to scavenge superoxide and peroxyl radicals. Also inhibits a variety of inflammatory responses. Reverses cardiac hypertrophy in mice via SIRT3 activation.

Chemical Identifiers

CAS 35354-74-6
Molecular Formula C18H18O2
Molecular Weight (g/mol) 266.34
InChI Key FVYXIJYOAGAUQK-UHFFFAOYSA-N
Synonym honokiol, 5,3'-diallyl-2,4'-dihydroxybiphenyl, 3,5'-diallyl-4,2'-dihydroxybiphenyl, unii-11513cco0n, chembl16901, 3',5-diallyl-1,1'-biphenyl-2,4'-diol, 4-allyl-2-3-allyl-4-hydroxy-phenyl phenol, 3',5-diallyl-2,4'-biphenyldiol, 3',5-diallylbiphenyl-2,4'-diol, 5,3′-diallyl-2,4′-dihydroxydiphenyl
PubChem CID 72303
ChEBI CHEBI:5759
IUPAC Name 2-(4-hydroxy-3-prop-2-enylphenyl)-4-prop-2-enylphenol
SMILES C=CCC1=CC(=C(C=C1)O)C2=CC(=C(C=C2)O)CC=C

Specifications

Quantity 10 mg
Formula Weight 266.33
Percent Purity >98%
Chemical Name or Material Honokiol
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.