Learn More
Medchemexpress LLC HOOC-PEG-SH MW 5000 50mg
SDP
Catalog No. 5000205101
Encompass_Preferred
HOOC-PEG-SH (MW 5000) (HOOC-PEG-Thiol (MW 5000)) is a reactive thiol PEG derivative with a terminal carboxyl group The carboxyl group can react with amine or hydroxyl groups to form a stable amide bond or an unstable ester bond The PEG linkage between the thiol and carboxyl groups has good water solubility flexible linker distance and higher stability[1]
Catalog No. 50-002-05101 Supplier Medchemexpress LLC Supplier No. HYW1048549C50MG
HOOC-PEG-SH (MW 5000) (HOOC-PEG-Thiol (MW 5000)) is a reactive thiol PEG derivative with a terminal carboxyl group The carboxyl group can react with amine or hydroxyl groups to form a stable amide bond or an unstable ester bond The PEG linkage between the thiol and carboxyl groups has good water solubility flexible linker distance and higher stability[1]