Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Hoocch2o-peg4-ch2cooh | 77855-75-5 | MFCD29079379 | >98.0% | 310.30 g/mol | C12H22O9 | 250MG
SDP

Catalog No. 5000236259
Encompass_Preferred

HOOCCH2O-PEG4-CH2COOH is a symmetric polyethylene glycol (PEG4) linker bearing terminal carboxylic acid groups. It is supplied as a high-purity research reagent intended for use as a spacer in PROTAC synthesis, bioconjugation, and linker optimization in medicinal chemistry. The reagent improves solubility relative to short alkyl linkers and provides a defined, flexible tether between functional moieties.

  • High purity ≥98.0%.
  • Molecular weight 310.30 g·mol-1.
  • Terminal carboxylic acids for straightforward conjugation chemistry.
  • PEG4 spacer provides defined length and hydrophilicity.
  • Soluble in DMSO for convenient handling.
  • Suitable for PROTAC synthesis and linker optimization studies.

Catalog No. 50-002-36259 Supplier Medchemexpress LLC Supplier No. HY124780250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

HOOCCH2O-PEG4-CH2COOH is a symmetric polyethylene glycol (PEG4) linker bearing terminal carboxylic acid groups. It is supplied as a high-purity research reagent intended for use as a spacer in PROTAC synthesis, bioconjugation, and linker optimization in medicinal chemistry. The reagent improves solubility relative to short alkyl linkers and provides a defined, flexible tether between functional moieties.

  • High purity ≥98.0%.
  • Molecular weight 310.30 g·mol-1.
  • Terminal carboxylic acids for straightforward conjugation chemistry.
  • PEG4 spacer provides defined length and hydrophilicity.
  • Soluble in DMSO for convenient handling.
  • Suitable for PROTAC synthesis and linker optimization studies.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.