Description
Premium suspension hoods and head covers
- Head covers provide head and face coverage; hoods cover head, face, and neck
- Fully assembled and ready to use
- Suitable for applications that require frequent replacement of the entire headgear assembly, such as pharmaceutical manufacturing, or situations where there is less soiling of the outer fabric
- Integrated models feature wide face seal with soft elastic for improved comfort and reduced noise
Premium Reusable Suspensions with Hoods
- Ideal for high disposal rate environments
- Soiled hood is replaced; suspension and air ducting are reused
- Rigid ducting for consistent air path
- Externally adjustable airflow control allows wearer to modify airflow distribution
- Suspension features independent adjustments for the crown and head circumference; allows custom fit for wide range of head sizes
- 3M Catalog# S-805-5, S-807-5, S-855, S-857
Specifications
Specifications
| Assigned Protection Factor (APF) | 1000 |
| Material | Zytron 200 |
| Product Type | Hood with Neck Collar |
| For Use With (Application) | Assembly, facility maintenance, machine operation, painting, sanding |
| For Use With (Equipment) | GVP, Breathe-Easy™, V Series, T-300, T-600, T-800 |
| Includes | S-805 hood with neck collar, S-950 Premium Head Suspension, S-955 Airflow Adjustment Cover, S-954 Size Reducing Pad, S-922 Peel-Off |
| Model | S-855 |
| Quantity | 1 Ea. |
| Suspension Type | Premium Resusable Suspension |
| Visor Type | Acetate |