Learn More
Cole-Parmer FHD-200 Standard Ductless Fume Hood, 36"W; 115 VAC, 60 Hz
Catalog No. NC1906082 Shop All Cole-Parmer Products
Encompass_Preferred
Ductless fume hoods are designed to be used with a wide range of applications to protect you and the environment. The hood is constructed of chemical-resistant epoxy-coated steel for added mechanical strength. All internal system components are isolated from fumes, extending the lifespan. LED task lights and clear acrylic side and ear panels allow ambient light through the chamber and provide you with a clear view of your contents. Combine one or more filter options to meet a wider range of multiple-use applications. Replacing the filter has never been easierno tools required! A filter access key is located at the front of the hood to prevent unauthorized removal or accidental exposure to dirty filters. Order filters, work tray, base stand, and other accessories separately to meet your application needs.
Catalog No. NC1906082 Supplier Cole-Parmer Supplier No. 7890002
Ductless fume hoods are designed to be used with a wide range of applications to protect you and the environment. The hood is constructed of chemical-resistant epoxy-coated steel for added mechanical strength. All internal system components are isolated from fumes, extending the lifespan. LED task lights and clear acrylic side and ear panels allow ambient light through the chamber and provide you with a clear view of your contents. Combine one or more filter options to meet a wider range of multiple-use applications. Replacing the filter has never been easierno tools required! A filter access key is located at the front of the hood to prevent unauthorized removal or accidental exposure to dirty filters. Order filters, work tray, base stand, and other accessories separately to meet your application needs.