Learn More
Med Vet International Smart Hoof Block Green Right, J0061XR, Each
Catalog No. 5000495884
Encompass_Preferred
The Smart Block. The use of hoof blocks adhered to the dairy's cow healthy claw has been a long standing practice. These blocks can often fall off too early, can be difficult to position properly and can be very uncomfortable for the cow. This can delay the healing process and prolong the lameness unnecessarily. The Smart Block is an innovative design with the hoof glue being channeled through an inlet port on the bottom to exit ports on top to evenly distribute the glue in the proper amount. Features/Benefits: - Saves Glue: Exact same amount of glue every time. No wasting glue, no air pickets, no guesswork. - Saves Time: The glue reaches the hoof and the block at exactly the same time eliminating any wasted curing time. Plus it ensures the most secure bond possible. - Soft but tough material gives flexibility and comfort durable enough to maintain its structure and shape throughout the healing process.
Catalog No. 50-004-95884 Supplier Med Vet International Supplier No. JOR61XR
The Smart Block. The use of hoof blocks adhered to the dairy's cow healthy claw has been a long standing practice. These blocks can often fall off too early, can be difficult to position properly and can be very uncomfortable for the cow. This can delay the healing process and prolong the lameness unnecessarily. The Smart Block is an innovative design with the hoof glue being channeled through an inlet port on the bottom to exit ports on top to evenly distribute the glue in the proper amount. Features/Benefits: - Saves Glue: Exact same amount of glue every time. No wasting glue, no air pickets, no guesswork. - Saves Time: The glue reaches the hoof and the block at exactly the same time eliminating any wasted curing time. Plus it ensures the most secure bond possible. - Soft but tough material gives flexibility and comfort durable enough to maintain its structure and shape throughout the healing process.