Learn More
MSC Vikan Pipe Exterior Brush, Medium Bristle, 2.2 in, 1 ea
Catalog No. 502869333
Encompass
This pipe exterior brush is engineered to maintain optimal hygiene by efficiently removing dust and contaminants from pipework. Its adjustable head design facilitates thorough cleaning of pipes up to 100 mm in outside diameter.
  • Adjustable head for cleaning pipes up to 100 mm outside diameter
  • Medium bristle stiffness
  • White bristle color
  • Constructed with polyester bristles, stainless steel, and polypropylene components
  • Complies with food contact and good manufacturing practice regulations (EC 1935/2004, EU 2023/2006/EC, FDA CFR 21, UK 2019 No. 704)
  • Meets REACH Regulation (EC) No. 1907/2006 and California Proposition 65
  • Halal and Kosher compliant
  • No intentionally added PFAS
Catalog No. 50-286-9333 Supplier MSC Supplier No. 50264514
This pipe exterior brush is engineered to maintain optimal hygiene by efficiently removing dust and contaminants from pipework. Its adjustable head design facilitates thorough cleaning of pipes up to 100 mm in outside diameter.
  • Adjustable head for cleaning pipes up to 100 mm outside diameter
  • Medium bristle stiffness
  • White bristle color
  • Constructed with polyester bristles, stainless steel, and polypropylene components
  • Complies with food contact and good manufacturing practice regulations (EC 1935/2004, EU 2023/2006/EC, FDA CFR 21, UK 2019 No. 704)
  • Meets REACH Regulation (EC) No. 1907/2006 and California Proposition 65
  • Halal and Kosher compliant
  • No intentionally added PFAS