Learn More
Chemglass Life Sciences Hook Connector, Stainless Steel
SDP
Supplier:  Chemglass Life Sciences CG9262S01
Encompass_Preferred
Hook Connector, Stainless Steel. Similar to CG-9262-01 except has a stainless steel electro-polished finish. Hook connector allows one-handed assembly of two components with one adjustment screw. Maximum diameter of 1/2" (13mm) for each component.
Catalog No. 50-200-730