Learn More
TrippNT 2 prong hook can replace or add hooks to your 24" or 48" adjustable surgical helmet rack. Made from non-metal, corrosion-resistant, and sterilizable HDPE, each hook supports up to 50 lbs for durable and flexible storage.
SDP
Catalog No. NC3356647 Shop All TrippNT Products
Encompass_Preferred
TrippNT's single 2 prong hook can be purchased to replace or add additional hooks to your 24 inch or 48 inch adjustable surgical helmet rack. Constructed from non-metal white high-density polyethylene (HDPE), this product is corrosion resistant and easily sterilized. Each hook has a 50 pound capacity.
Catalog No. NC3356647 Supplier TrippNT Supplier No. 53942
TrippNT's single 2 prong hook can be purchased to replace or add additional hooks to your 24 inch or 48 inch adjustable surgical helmet rack. Constructed from non-metal white high-density polyethylene (HDPE), this product is corrosion resistant and easily sterilized. Each hook has a 50 pound capacity.