Learn More
Med Vet International 8-in (20Cm) Snook Hook, Spay & Neuter, 4-5mm Width
Catalog No. 502223941
Encompass_Preferred
Snook hooks are surgical instrument with a curved, button tip used to retrieve a horn of the uterus when performing ovariohysterectomy in dogs and cats.
Catalog No. 50-222-3941 Supplier Med Vet International Supplier No. 1101
Snook hooks are surgical instrument with a curved, button tip used to retrieve a horn of the uterus when performing ovariohysterectomy in dogs and cats.