Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hook-Up Wire
SCI_ED

Catalog No. S48310
Change view
Click to view available options
Red
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
S48310 Red
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S48310 Supplier Sempco Inc Supplier No. WS0192

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Functional and versatile for all your needs.

  • 18-gauge copper wire
  • Insulated with a plastic coating
  • Length: 50 ft. (15.24m)

Specifications

Product Type Hook-up Wire
Length (English) 50 ft.
Length (Metric) 15.24 m
Material Copper, Plastic
Color Red
Gauge 18 ga
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.