Learn More
ABclonal Technology HOOK3 Rabbit pAb
SDP
Supplier:  ABclonal Technology A155361000UL
Encompass_Preferred
Hook proteins are cytosolic coiled-coil proteins that contain conserved N-terminal domains, which attach to microtubules, and more divergent C-terminal domains, which mediate binding to organelles. The Drosophila Hook protein is a component of the endocytic compartment.
Catalog No. 50-001-05833