Learn More
Supertek™ Hooke's Law Kit with Activity Sheet
GREENER_CHOICESCI_ED
Catalog No. S01261 Shop All Supertek Scientific Products
Change view
Click to view available options
Hooke's Law Kit
Catalog No.
S01261 Hooke's Law Kit
1 options
Catalog No. S01261 Supplier Supertek™ Supplier No. PH10612AS3
Add to Cart
Add to Cart
This Hooke’s Law Kit shows you how to make a scale to weigh many household objects using Hooke’s Law.
  • This Hooke's Law Kit consists of 1 tape measure, a set of colorful plastic hex weights, a highly accurate special spring and a detailed activity guide
  • Ages 7 plus
Specifications
Product Type Hooke's Law Kit