Learn More
Med Vet International 8in (20cm) Spay & Neuter Snook Hook, 5-6mm Wide, German, Von Klaus
Supplier:  Med Vet International VK1201620
Encompass_Preferred
A Snook hook is a surgical instrument with a curved, button tip used to retrieve a horn of the uterus when performing ovariohysterectomy in dogs and cats. The Von Klaus line of instruments is made of high quality German stainless steel, expertly crafted and comes with a lifetime warranty when used for their intended surgical purpose.
Catalog No. 50-209-2285
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.