Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hop-22(29)-ene Solution, 0.1 mg/mL in Isooctane, ≥85% (GC/MS), MilliporeSigma™ Supelco™

Catalog No. 111017995 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-7995 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-7995 Supplier MilliporeSigma Supelco Supplier No. 046261ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Certan Vial

Food and Beverages
TRUSTED_SUSTAINABILITY

Specifications

CAS 1615-91-4
Flash Point -12°C
Linear Formula C30H50
MDL Number MFCD00216319
Synonym Diploptene
Type Analytical Standard
Quantity 1 mL
Formula Weight 410.72
Percent Purity ≥85% (GC/MS)
Grade Analytical Standard
Concentration or Composition (by Analyte or Components) 0.1 mg/mL in isooctane
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.