Learn More
Enzo Life Sciences HOP (human), (recombinant) (200µg)
SDP
Supplier:  Enzo Life Sciences ADISRP1510F
Encompass_Preferred
Alternative name: HSC70/HSP90-organizing protein, STIP1, STI1, Stress-induced phosphoprotein 1, STUB1, p60, CHIP. Purity: ≥90% (SDS-PAGE; Western blot). MW: ~63kDa. Formulation: Liquid. In Dulbecco's PBS. Source: Produced in E. coli. Long Term Storage: -80°C. Applications: WB .
Catalog No. 50-200-7676