Learn More
Enzo Life Sciences HOP monoclonal antibody (DS14F5) (200µg)
SDP
Supplier:  Enzo Life Sciences ADISRA1500F
Encompass_Preferred
Clone: DS14F5. Host: Mouse. Isotype: IgG1. Immunogen: Chicken HOP (p60). UniProt ID: P31948. Source: Purified from mouse ascites. Species reactivity: Human, Mouse, Rat, Xenopus, Sheep, Rabbit, Porcine, Monkey, Mink, Hamster, Guinea pig, Dog, Chicken, Bovine. Applications: IHC, IP, WB. Recommended Dilutions: Western Blot (1:1,000, colorimetric) Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Protein G affinity purified. Formulation: Liquid. In PBS, pH 7.2, containing 50% glycerol and 0.09% sodium azide. Handling: Avoid freeze/thaw cycles.
Catalog No. 50-200-7673