Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medicom™ Hopen™ Anti-Static Hygiene Gown

Catalog No. 19910577 Shop All Medicom Products
Change view
Click to view available options
Size:
Large
X-Large
2X-Large
3X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-910-577 Large
19-910-578 X-Large
19-910-579 2X-Large
19-910-580 3X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 19-910-577 Supplier Medicom™ Supplier No. 40.507/03
This item is not returnable. View return policy

Medicom™ Hopen™ Anti-Static Hygiene Gown with a waterproof, microporous polyethylene lining

  • Single use, anti-static hygiene gown provides the user with full body protection
  • Protects against activity related soiling and contamination
  • Microporous, polyethylene lining is comfortable, and waterproof

Specifications

Packaging Type Individual, Bag
Product Type Hygiene Gown
Size Large
For Use With (Application) Category 1 Personal Protective Equipment
Gender Unisex
No. of Pockets None
Certifications/Compliance (Europe) EN 1149-5
Construction Made in China
Material Polypropylene, SMS, Microporous Polyethylene Laminate
Color White
Closure Type Zipper
Disposable Single-use
Sterility Non-sterile
Cuff Style Elastic
Seam Type Sewn
Certifications/Compliance Manufactured under an ISO 9001 Certified Quality System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.