Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medicom™ Hopen™ Boot Covers

Catalog No. 19910692 Shop All Medicom Products
Change view
Click to view available options
Thickness:
40 g/m2
55 g/m2
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Thickness
19-910-692 40 g/m2
19-910-691 55 g/m2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-910-692 Supplier Medicom™ Supplier No. 10.25

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This item is not returnable. View return policy

Medicom™ Hopen™ Boot Covers for use in ISO 6, 7, and 8 environment types or the food industry

  • Recommened for use in the fields of food and protected environments
  • Porous material allows for better breathability
  • PVC sole is completely waterproof, and ultra resistant to high pressure cleaning
  • Can be used indoors or outdoors
  • Fully covers the entire boot

Specifications

Size Universal
Material Polypropylene
Color White
Disposable Single-use
Sterility Non-sterile
Closure Type Elastic Top
Thickness 40 g/m2
For Use With (Application) ISO 6, 7, 8 Environment Types
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.