Learn More
Medicom™ Hopen™ HOP'SAFE Hooded Coveralls
Catalog No. 19910585 Shop All Medicom Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-910-585 Small
19-910-584 Medium
19-910-583 Large
19-910-586 X-Large
19-910-587 2X-Large
19-910-588 3X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 19-910-585 Supplier Medicom™ Supplier No. 40.040/01
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
This item is not returnable. View return policy
Medicom™ Hopen™ HOP'SAFE Hooded Coveralls are anti-static, and protect against radioactive particles
  • These HOP'SAFE coveralls are hooded with an adhesive flap on the zipper closure
  • Polypropylene SMS material is air and water vapor permeable
  • Coverall EPI, Category III, Type 5/6
  • External seams provide additional comfort
  • Good tear, and abrasion resistance
  • Single use, non sterile
  • Individually packaged
Specifications
Product Type Single-use Coveralls
Size Small
Gender Unisex
Material Polypropylene, SMS
Color White
Disposable Single-use
Waist Style Elastic
Ankle Style Elastic
Cuff Style Elastic
Neck Style Hooded
Closure Type Front Zipper
Sterility Non-sterile
Resistance Anti-static, Particle
Certifications/Compliance (Europe) EN1149-5, EN1073-2
No. of Pockets None
Thickness 55 g/m2