Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medicom™ Hopen™ HOP'SAFE PRO Coveralls

Catalog No. 19910593 Shop All Medicom Products
Change view
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
3X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-910-593 3X-Large
19-910-589 Medium
19-910-590 Large
19-910-591 X-Large
19-910-592 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 19-910-593 Supplier Medicom™ Supplier No. 40.035/06
This item is not returnable. View return policy

Medicom™ Hopen™ HOP'SAFE PRO Coveralls are extremely resistant and offer users maximum protection

  • HOP'SAFE PRO Coveralls offer Category I protection
  • Lint-free material is tear and abrasion resistant
  • Offer users maximum protection

Specifications

Product Type Single-use Coveralls
Size 3X-Large
Gender Unisex
Material Polypropylene, SMS
Color Blue
Disposable Single-use
Ankle Style Elastic
Cuff Style Thumb Loop
Neck Style Hooded
Hood Type Elastic
Sterility Non-sterile
No. of Pockets None
Thickness 50 g/m2
For Use With (Application) Category I Personal Protective Equipment
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.