Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medicom™ Hopen™ Hygiene Eco Coveralls

Catalog No. 19910617 Shop All Medicom Products
Change view
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-910-617 Medium
19-910-618 Large
19-910-619 X-Large
19-910-620 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 19-910-617 Supplier Medicom™ Supplier No. 40.103/02
This item is not returnable. View return policy

Medicom™ Hopen™ Hygiene Eco Coveralls are suitable to HACCP procedures

  • These Hygiene Eco white coveralls protect the working environment from particles coming from the body of the operator
  • Allows the operator to protect himself from the risks of soiling related to the activity being carried out
  • Category 1 Personal Protective Equipment
  • Suitable to HACCP procedures

Specifications

Product Type Single-use Coveralls
Size Medium
Gender Unisex
Material Polypropylene
Color White
Disposable Single-use
Ankle Style Elastic
Cuff Style Elastic
Neck Style Collared
Closure Type Zipper
Seam Type Stitched
Sterility Non-sterile
No. of Pockets None
For Use With (Application) Category 1 Personal Protective Equipment
Weight (Metric) 30 g/m2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.